accessibility menu, dialog, popup

UserName

CH3NOO

CAS RN 3222-47-7

IUPAC Name: 6-methylpyridine-3-carboxylate
Synonyms: 6-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H6NO2
InChi Key: RZOKQIPOABEQAM-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=N1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results