accessibility menu, dialog, popup

UserName

CAS RN 3152-12-3

HOOOHClCl

CAS RN 3152-12-3

IUPAC Name: 2,2-bis(2-chlorophenyl)-2-hydroxyacetic acid
Synonyms: bis(2-chlorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol): 297.13
Molecular Formula: C14H10Cl2O3
InChi Key: OJYHUFNZIWRMPT-UHFFFAOYSA-N
SMILES: OC(=O)C(O)(C1=CC=CC=C1Cl)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results