missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichlorobenzilic acid, 98+%, Thermo Scientific™

Product Code. 11342486
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11342486

Brand: Thermo Scientific Alfa Aesar L06356.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3152-12-3
MDL Number MFCD00049223

Specifications

Melting Point ∼162°C
Molecular Formula C14H10Cl2O3
Quantity 10 g
Beilstein 2387319
Synonym 2,2'-dichlorobenzilic acid, 2,2-bis 2-chlorophenyl-2-hydroxyacetic acid, bis 2-chlorophenyl hydroxy acetic acid, 4,5'-sulfonyldiphenol, acmc-1cupg, 2,2'-dichlorobrnzilic acid, 2,2'-dichlorobenzilic acid, 98+%, bis 2-chlorophenyl hydroxyacetic acid, bis-2-chloro-phenyl-hydroxy-acetic acid
InChI Key OJYHUFNZIWRMPT-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(C2=CC=CC=C2Cl)(C(=O)O)O)Cl
IUPAC Name 2,2-bis(2-chlorophenyl)-2-hydroxyacetic acid
Molecular Weight (g/mol) 297.13
PubChem CID 329060
Formula Weight 297.14
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dichlorobenzilic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.