accessibility menu, dialog, popup

UserName

H3CCH3HNNHONOHPOHOHO

CAS RN 305-33-9

IUPAC Name: N'-(propan-2-yl)pyridine-4-carbohydrazide; phosphoric acid
Synonyms: iproniazid; phosphoric acid
Molecular Weight (g/mol): 277.22
Molecular Formula: C9H16N3O5P
InChi Key: YPDVTKJXVHYWFY-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.CC(C)NNC(=O)C1=CC=NC=C1
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results