Learn More
Iproniazid Phosphate, LoGiCal
Product Code. 30464121
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464121 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30464121 Supplier LoGiCal Supplier No. LGCFOR1107.00
 Restricted Item
Estimated Shipment: 02-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iproniazid Phosphate
CAS 305-33-9
Analyte or Component Names Iproniazid Phosphate
Shipping Condition Room Temperature
Molecular Formula C9 H13 N3 O . H3 O4 P
Certifications/Compliance ISO 9001
Synonym 4-Pyridinecarboxylic acid, 2-(1-methylethyl)hydrazide, phosphate (1:1), Isonicotinic acid, 2-isopropylhydrazide, phosphate (1:1) (8CI), 1-Isonicotinyl-2-isopropylhydrazine phosphate, 1-Isopropyl-2-isonicotinylhydrazine phosphate, Iproniazid dihydrogen phosphate, Iproniazid phosphate, Iproniazide phosphate, Marsilid phosphate, NSC 81850
InChI Formula InChI=1S/C9H13N3O.H3O4P/c1-7(2)11-12-9(13)8-3-5-10-6-4-8;1-5(2,3)4/h3-7,11H,1-2H3,(H,12,13);(H3,1,2,3,4)
Solution Type Neat
SMILES CC(C)NNC(=O)c1ccncc1.OP(=O)(O)O
IUPAC Name phosphoric acid;N'-propan-2-ylpyridine-4-carbohydrazide
Industry Type Clinical Testing
Molecular Weight (g/mol) 277.21
Quantity 10 mg
Formula Weight 277.0828 g/mol