Learn More
Iproniazid Phosphate, Mikromol™
Description
Iproniazid Phosphate
Specifications
Specifications
| Chemical Name or Material | Iproniazid Phosphate |
| CAS | 305-33-9 |
| Analyte or Component Names | Iproniazid Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N3 O . H3 O4 P |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Pyridinecarboxylic acid, 2-(1-methylethyl)hydrazide, phosphate (1:1), Isonicotinic acid, 2-isopropylhydrazide, phosphate (1:1) (8CI), 1-Isonicotinyl-2-isopropylhydrazine phosphate, 1-Isopropyl-2-isonicotinylhydrazine phosphate, Iproniazid dihydrogen phosphate, Iproniazid phosphate, Iproniazide phosphate, Marsilid phosphate, NSC 81850 |
| InChI Formula | InChI=1S/C9H13N3O.H3O4P/c1-7(2)11-12-9(13)8-3-5-10-6-4-8;1-5(2,3)4/h3-7,11H,1-2H3,(H,12,13);(H3,1,2,3,4) |
| SMILES | CC(C)NNC(=O)c1ccncc1.OP(=O)(O)O |
| IUPAC Name | phosphoric acid;N'-propan-2-ylpyridine-4-carbohydrazide |
| Show More |