accessibility menu, dialog, popup

UserName

CAS RN 109-08-0

CH3NN

CAS RN 109-08-0

IUPAC Name: 2-methylpyrazine
Synonyms: 2-methylpyrazine
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: CAWHJQAVHZEVTJ-UHFFFAOYSA-N
SMILES: CC1=CN=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)