missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZW10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
391.65€ - 590.10€
Specifications
| Antigen | ZW10 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600166
|
Novus Biologicals
NBP2-38751-25ul |
25 μL |
415.00€ 391.65€ / 25µL Save 23.35€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18153869
|
Novus Biologicals
NBP2-38751 |
0.1 mL |
624.00€ 590.10€ / 0.10mL Save 33.90€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZW10 Polyclonal specifically detects ZW10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZW10 | |
| Polyclonal | |
| Rabbit | |
| Mitotic Regulators | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| E3 ubiquitin-protein ligase ZSWIM2, EC 6.3.2.-, MEKK1-related protein X, MEX, MGC33890, Zinc finger SWIM domain-containing protein 2, zinc finger, SWIM domain containing 2, zinc finger, SWIM-type containing 2, ZZZ2ZZ-type zinc finger-containing protein 2 | |
| ZW10 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O43264 | |
| 9183 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KGEVCNMISKKYSEFLPSMQSAQGLITQVDKLSEDIDLLKSRIESEVRRDLHVSTGEFTDLKQQLERDSVVLSLLKQLQEFSTAIEEYNCALTEKKYVTG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title