missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZSCAN5 Antibody (PCRP-ZSCAN5A-2H4), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | ZSCAN5 |
|---|---|
| Clone | PCRP-ZSCAN5A-2H4 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043431
|
Novus Biologicals™
NBP3-26750-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043197
|
Novus Biologicals™
NBP3-26750-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZSCAN5 Monoclonal antibody specifically detects ZSCAN5 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZSCAN5 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495 | |
| Recombinant fragment (around aa37-124) of human ZSCAN5 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-ZSCAN5A-2H4 | |
| Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 79149 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title