missing translation for 'onlineSavingsMsg'
Learn More

ZNF638 Antibody, Novus Biologicals™

Product Code. 18284050 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18284050 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18284050 Supplier Novus Biologicals Supplier No. NB10077290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF638 Polyclonal specifically detects ZNF638 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF638
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation (Negative)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_055312.2
Gene Alias CTCL tumor antigen se33-1, CTCL-associated antigen se33-1, Cutaneous T-cell lymphoma-associated antigen se33-1, DKFZp686P1231, MGC26130, MGC90196, NP220 nuclear protein, NP220zinc finger, matrin-like, Nuclear protein 220, ZFML, Zfp638, Zinc finger matrin-like protein, zinc finger protein 638
Gene Symbols ZNF638
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human zinc finger protein 638 (Nuclear protein 220) using the numbering given in entry NP_055312.2 (GeneID 27332).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27332
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.