missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF622 Mouse anti-Human, Clone: PCRP-ZNF622-1C11, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | ZNF622 |
|---|---|
| Clone | PCRP-ZNF622-1C11 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Beschreibung
ZNF622 Monoclonal specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Spezifikation
| ZNF622 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
| Recombinant full-length human ZNF622 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-ZNF622-1C11 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 90441 | |
| IgG2a | |
| Protein A or G purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts