missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF620 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94038-0.1ml
This item is not returnable.
View return policy
Description
ZNF620 Polyclonal antibody specifically detects ZNF620 in Mouse samples. It is validated for Western Blot
Specifications
| ZNF620 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| zinc finger protein 620 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human ZNF620 (NP_001243096.1). MVGGLPGNVSQHLDFGSSLEQPQGHWIIKTKSKRRHFTDTSARHHEAYEVKNGEKFEKLGKNISVSTQLTTNQTNPSGQISYECGQCGRYFIQMADFHRHEKCHTGEKSFECKECGKYFRYNSLLIRHQIIHTGKKPFKCKECGKGLSSD | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 253639 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction