Learn More
ZNF619 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | ZNF619 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF619 Polyclonal specifically detects ZNF619 in Human samples. It is validated for Western Blot.Specifications
| ZNF619 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 285267 | |
| Synthetic peptide directed towards the N terminal of human ZNF619. Peptide sequence: RLIVEGLLMDVPQHPDFKDRLEKSQLHDTGNKTKIGDCTDLTVQDHESST | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ90764, zinc finger protein 619 | |
| ZNF619 | |
| IgG | |
| 63 kDa |