missing translation for 'onlineSavingsMsg'
Learn More

ZNF600 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16829584 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16829584 150 μL 150µL
16839584 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16829584 Supplier Proteintech Supplier No. 201001AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is specific to ZNF600.

Consisting of around 63million baseswith over 1,400 genes,chromosome 19 makes up over 2% of human genomic DNA. Chromosome 19 includes a diversity of interesting genes and isrecognized for having the greatest gene density of the human chromosomes. It isthe genetic home for a number of immunoglobulin superfamily members including the killercell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAMand PSG family, and Fcalpha receptors. Key genesfor eye color and haircolor also map to chromosome 19. Peutz-Jegherssyndrome,spinocerebellar ataxia type 6, the stroke disorder CADASIL, hypercholesterolemia and Insulin-dependent diabetes have been linked to chromosome 19. Translocations with chromosome 19 and chromosome 14 can be seen in some lymphoproliferative disorders and typicallyinvolve the proto-oncogene Bcl-3.

Specifications

Antigen ZNF600
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZNF600
Gene Accession No. Q6ZNG1
Gene Alias KR ZNF1, zinc finger protein 600, ZNF600
Gene Symbols Zfp600, ZNF600
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162966, 667666
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.