missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF483 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | ZNF483 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883815
|
Proteintech
25242-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16873815
|
Proteintech
25242-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF483 may be involved in transcriptional regulation.Specifications
| ZNF483 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8TF39 | |
| 158399 | |
| ZNF483 Fusion Protein Ag18739 | |
| Primary | |
| -20°C | |
| ZNF483 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| KIAA1962, zinc finger protein 483, ZKSCAN16, ZNF483 | |
| ZNF483 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title