missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF440 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | ZNF440 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF440 Polyclonal specifically detects ZNF440 in Human samples. It is validated for Western Blot.Specifications
| ZNF440 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ37933, zinc finger protein 440 | |
| ZNF440 | |
| IgG | |
| 69 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_689570 | |
| 126070 | |
| Synthetic peptide directed towards the middle region of human ZNF440. Peptide sequence ECGKAFTCPRYVRIHERTHSRKNLYECKQCGKALSSLTSFQTHVRLHSGE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title