missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF329 Antibody (OTI9F9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | ZNF329 |
|---|---|
| Clone | OTI9F9 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306714
|
Novus Biologicals
NBP3-44970-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306376
|
Novus Biologicals
NBP3-44970-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF329 Monoclonal antibody specifically detects ZNF329 in Human samples. It is validated for Western BlotSpecifications
| ZNF329 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 79673 | |
| IgG2a | |
| >95% | |
| Antibody |
| OTI9F9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| zinc finger protein 394, zinc finger protein 99, Zinc finger protein with KRAB and SCAN domains 14, ZKSCAN14FLJ12298 | |
| Human recombinant protein fragment corresponding to amino acids 1-203 of human ZNF329 (NP_078896) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title