missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF286 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80345
This item is not returnable.
View return policy
Description
ZNF286 Polyclonal specifically detects ZNF286 in Human samples. It is validated for Western Blot.
Specifications
| ZNF286 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC149627, zinc finger protein 286, zinc finger protein 286A | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 57335 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_065703 | |
| ZNF286A | |
| Synthetic peptide directed towards the C terminal of human ZNF286. Peptide sequence GKAFIHSSALIQHQRTHTGEKPFRCNECGKSFKCSSSLIRHQRVHTEEQP. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Equine: 92%; Rabbit: 92%; Bovine: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction