Learn More
ZNF281 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24088-1-AP-150UL
This item is not returnable.
View return policy
Description
The ZNF281 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNF281. This antibody recognizes human antigen. The ZNF281 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| ZNF281 | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| GZP1, Transcription factor ZBP 99, ZBP 99, ZBP99, zinc finger protein 281, ZNF281, ZNP 99 | |
| 150 μL | |
| 23528 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |