missing translation for 'onlineSavingsMsg'
Learn More

ZNF281 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76551-0.025mg

 View more versions of this product

Product Code. 18582321

  • 178.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF281 Polyclonal specifically detects ZNF281 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF281
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
FLJ20557, zinc finger protein 416
Rabbit
Affinity Purified
RUO
23528
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL
AAH51905
ZNF281
Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ZFP281. The immunogen is located within amino acids 600 - 650 of ZFP281. Amino Acid Squence: YSNKSESQKEDPFNIAEPR
0.025 mg
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only