missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF263 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00€ - 584.00€
Specifications
| Antigen | ZNF263 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231383
|
Novus Biologicals
NBP3-35673-20ul |
20 μL |
214.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232450
|
Novus Biologicals
NBP3-35673-100ul |
100 μL |
584.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF263 Polyclonal antibody specifically detects ZNF263 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| ZNF263 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| GIOT-1GIOT1MGC33911, gonadotropin inducible transcription repressor 1, Gonadotropin-inducible ovary transcription repressor 1, zinc finger protein 461 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-240 of human ZNF263 (NP_005732.2).,, Sequence:, QRELGRLRQQVTNHGRGTEVLLEEPLPLETARESPSFKLEPMETERSPGPRLQELLGPSPQRDPQAVKERALSAPWLSLFPPEGNMEDKEMTGPQLPESLEDVAMYISQEEWGHQDPSKRA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10127 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title