missing translation for 'onlineSavingsMsg'
Learn More

ZNF239 Antibody (PCRP-ZNF239-2A10), Allophycocyanin, Novus Biologicals™

Product Code. 18603934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18603934 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18603934 Supplier Novus Biologicals Supplier No. NBP324148APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF239 Monoclonal antibody specifically detects ZNF239 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF239
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-ZNF239-2A10
Conjugate APC
Formulation PBS
Gene Alias KIAA1198, zinc finger protein 490
Host Species Mouse
Immunogen Recombinant fragment (around aa83-233) of human ZNF239 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8187
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.