missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF227 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
484.00€
Specifications
| Antigen | ZNF227 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
ZNF227 Polyclonal specifically detects ZNF227 in Human samples. It is validated for Western Blot.Specifications
| ZNF227 | |
| Polyclonal | |
| Purified | |
| RUO | |
| AAO45835 | |
| 7770 | |
| Synthetic peptide directed towards the middle region of human ZNF227. Peptide sequence ESGDCFNKSSFHSYQSNHTGEKSYRCDSCGKGFSSSTGLIIHYRTHTGEK. | |
| Primary | |
| 30 kDa |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KIAA1084, zinc finger protein 507 | |
| ZNF227 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title