missing translation for 'onlineSavingsMsg'
Learn More

ZNF200 Monoclonal Antibody (OTI3B6), TrueMAB™, OriGene

Product Code. 17599203
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17599203 30 μL 30µL
17579203 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17599203 Supplier Origene Technologies Supplier No. TA808210S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF200 Monoclonal antibody specifically detects ZNF200 in Human samples. It is validated for Western Blot

Specifications

Antigen ZNF200
Applications Western Blot
Classification Monoclonal
Clone OTI3B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZNF200
Gene Accession No. P98182
Gene Alias MGC45293; Zinc finger protein 200; ZN200; ZNF200; ZNFMF
Host Species Mouse
Immunogen Full length human recombinant protein of human ZNF200 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7752
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.