missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94653-0.1ml
This item is not returnable.
View return policy
Description
ZNF12 Polyclonal antibody specifically detects ZNF12 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZNF12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| GIOT3, Gonadotropin-inducible ovary transcription repressor 3, HZF11, zinc finger protein 11, zinc finger protein 12, Zinc finger protein 325GIOT-3KOX3gonadotropin inducible transcription repressor 3, Zinc finger protein KOX3, ZNF325 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-240 of human ZNF12 (NP_057349.2). EGEFLLQSYPDEVWQTDDLIERIQEEENKPSRQTVFIETLIEERGNVPGKTFDVETNPVPSRKIAYKNSLCDSCEKCLTSVSEYISSDGSYARMKADECSGCGKSLLHIKLEKTHPGDQAYEFNQNGEPYTLNEESLYQKIRILEKPFEYIECQKAFQKDTVFVNHMEEKP | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7559 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction