missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76525-0.025mg
This item is not returnable.
View return policy
Description
ZMYM3 Polyclonal specifically detects ZMYM3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ZMYM3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of ZMYM3 are known to exist; this antibody will detect all three. ZMYM3 antibody is predicted to not cross-react with other ZMYM protein family members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| AAH69057 | |
| ZMYM3 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the center of human ZMYM3. The immunogen is located within amino acids 220 - 270 of ZMYM3. | |
| 0.025 mg | |
| DNA Repair | |
| 9203 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction