missing translation for 'onlineSavingsMsg'
Learn More

Zinc finger protein 774 Antibody (PCRP-ZNF774-3F7) - Azide and BSA Free, Novus Biologicals™

Product Code. 18674505 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18674505 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18674505 Supplier Novus Biologicals Supplier No. NBP320746

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zinc finger protein 774 Monoclonal antibody specifically detects Zinc finger protein 774 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zinc finger protein 774
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone PCRP-ZNF774-3F7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-2 μg/million cells, ELISA
Formulation 10mM PBS
Gene Alias MGC75360, zinc finger protein 774
Host Species Mouse
Immunogen Recombinant full-length human Zinc finger protein 774 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342132
Target Species Human
Content And Storage Store at -20°C to -70°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.