missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) NS1 Antibody (10), Novus Biologicals™
Mouse Monoclonal Antibody
241.00€ - 448.00€
Specifications
| Antigen | Zika Virus (SPH2015) NS1 |
|---|---|
| Clone | 10 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | Sandwich ELISA Detection |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625556
|
Novus Biologicals
NBP3-05858-100ul |
100 μg |
448.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609906
|
Novus Biologicals
NBP3-05858-50ul |
50 μg |
241.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zika Virus (SPH2015) NS1 Monoclonal antibody specifically detects Zika Virus (SPH2015) NS1 in Virus samples. It is validated for Sandwich ELISA DetectionSpecifications
| Zika Virus (SPH2015) NS1 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val796-Leu1157). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 10 | |
| Sandwich ELISA Detection | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Zika NS1, ZIKV NS1, ZIKV-NS1 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title