Learn More
Invitrogen™ Zika Virus NS1 (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (201)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560382
This item is not returnable.
View return policy
Description
Zika Virus NS1 (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. & Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
Specifications
| Zika Virus NS1 (strain Zika SPH2015) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA | |
| 201 | |
| Unconjugated | |
| Zika NS1; ZIKV NS1; ZIKV-NS1 | |
| Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 protein (Val796-Leu1157). | |
| 20 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |