missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ L1TD1 Monoclonal Antibody (901258)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561937

Product Code. 30536105

  • 469.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

L1TD1 is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

L1TD1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q5T7N2
L1TD1
E. coli-derived recombinant human L1TD1, Met1-Leu142.
100 μg
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
901258
Unconjugated
L1TD1
ECAT11; ES cell-associated protein 11; L1TD1; LINE1 type transposase domain containing 1; LINE-1 type transposase domain containing 1; LINE-1 type transposase domain-containing protein 1; RP5-1155K23.3
Mouse
Protein A/G
RUO
54596
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.