missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4B7/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 351.00€
Specifications
| Antigen | Zebrafish Gut Absorptive Cell Epitopes |
|---|---|
| Clone | FIS 4B7/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602008
|
Novus Biologicals
NBP2-50373-0.025ml |
0.025 mL |
190.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663939
|
Novus Biologicals
NBP2-50373 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zebrafish Gut Absorptive Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Zebrafish Gut Absorptive Cell Epitopes | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| PBS | |
| IgG1 | |
| Protein A purified |
| FIS 4B7/1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Zebrafish | |
| Zebrafish intestine | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title