missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB1 Antibody (CL0151) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52866PE
This item is not returnable.
View return policy
Description
ZEB1 Monoclonal antibody specifically detects ZEB1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| ZEB1 | |
| Monoclonal | |
| PE | |
| AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
| This antibody was developed using a recombinant protein derived from P37275, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger | |
| 6935 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0151 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction