missing translation for 'onlineSavingsMsg'
Learn More

ZDHHC5 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18658572 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18658572 0.02 mL 0.02mL
18604602 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18658572 Supplier Novus Biologicals Supplier No. NBP2944700.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZDHHC5 Polyclonal antibody specifically detects ZDHHC5 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias DHHC domain containing 5, DKFZp586K0524, zinc finger, DHHC-type containing 5, ZNF375
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 130-230 of human ZDHHC5 (NP_056272.2). FDHHCPWVNNCIGRRNYRYFFLFLLSLTAHIMGVFGFGLLYVLYHIEELSGVRTAVTMAVMCVAGLFFIPVAGLTGFHVVLVARGRTTNEQVTGKFRGGVN
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 25921
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.