missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H7A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00€ - 470.00€
Specifications
| Antigen | ZC3H7A |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611391
|
Novus Biologicals
NBP2-93195-0.02ml |
0.02 mL |
193.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633770
|
Novus Biologicals
NBP2-93195-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZC3H7A Polyclonal antibody specifically detects ZC3H7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZC3H7A | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 29066 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ10027, FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH domain-containing protein 7A, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type containing 7A, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZC3H7A (NP_054872.2). MSNVSEERRKRQQNIKEGLQFIQSPLSYPGTQEQYAVYLRALVRNLFNEGNDVYREHDWNNSISQYTEALNIADYAKSEEILIPKEIIEKLYINRIACYSNMGFHDKVLEDCNIVLSLNASNCKALYRKSKALSDLGRYKKAYDAVAKCSLAVPQDEHVIKLTQELAQKLGFKIRKAYVRAELSLKSVPGDGATKALNHSVEDIEPDLLTPRQEAVPVVS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title