missing translation for 'onlineSavingsMsg'
Learn More

ZBED2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18782954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.03mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782954 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18782954 Supplier Novus Biologicals Supplier No. NBP1773490.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBED2 Polyclonal specifically detects ZBED2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_078784
Gene Alias MGC10796, zinc finger BED domain-containing protein 2, zinc finger, BED domain containing 2, zinc finger, BED-type containing 2
Gene Symbols ZBED2
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human ZBED2. The immunogen is located within the first 50 amino acids of ZBED2. Amino Acid Squence: DLEMKEEEEISETGE
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79413
Test Specificity This antibody is predicted to have no cross-reactivity to ZBED1 or ZBED3.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.