Learn More
MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
Description
Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).
Specifications
Specifications
| Color | White |
| Content And Storage | 4°C |
| Quantity | 3 mg |
| Product Type | Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor |
| Molecular Weight (g/mol) | 467.487 |
| Form | Solid |