Learn More
Novus Biologicals™ Z-LE(OMe)HD(OMe)-FMK (Caspase 9 Inhibitor)
Shop All Bio Techne Products
Click to view available options
Quantity:
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Color | Tan |
| For Use With (Application) | In vitro assay |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles |
| Quantity | 5 mg |
| Product Type | Z-LE(OMe)HD(OMe)-FMK (Caspase 9) Inhibitor |
| Molecular Weight (g/mol) | 690 |
| Inhibitors | Z-LE(OMe)HD(OMe)-FMK (Caspase 9) |
| Form | Crystalline Powder |