missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone

Product Code. p-8015229 Shop All MP Biomedical Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 11434906

Brand: MP Biomedicals™ 03FK01001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-FMK is an irreversible and cell permeable inhibitor of CPP32/Apopain.

A member of the CIE/CED-3 family of cysteine proteases.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store in a cool, dry place. Dessicate.
Quantity 1 mg
Product Type Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone
Molecular Weight (g/mol) 668
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.