missing translation for 'onlineSavingsMsg'
Learn More

Abcam (Z)-4-Hydroxytamoxifen, Estrogen Receptor Modulator, Solid, 25 mg

Product Code. 32117694
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32117694 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32117694 Supplier Abcam Supplier No. ab14194325mg
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective estrogen receptor antagonist with higher potency in vitro than (Z)-tamoxifen.

  • Molecular Weight: 387.5 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Potent and selective estrogen receptor antagonist (IC50 = 7 nM); active metabolite of (Z)-tamoxifen (ab120656) with higher in vitro potency
  • Biological Effect: Shows antiproliferative effect (IC50 = 27 μM) and induces p21 nuclear localization
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 4-Hydroxytamoxifen
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Target Molecule Estrogen Receptor
Formulation 100% 4-Hydroxytamoxifen
Quantity 25 mg
For Use With (Application) Estrogen receptor modulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.