missing translation for 'onlineSavingsMsg'
Learn More

YWHAZ Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen YWHAZ
Clone 3G12B10
Concentration 700 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236121
View Documents
Proteintech
67698-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234300
View Documents
Proteintech
67698-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The YWHAZ antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YWHAZ. This antibody recognizes human, mouse, rat antigen. The YWHAZ antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications

Specifications

YWHAZ
700 μg/mL
Monoclonal
Liquid
RUO
7534
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
3G12B10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Mouse
Human, Mouse, Rat
KCIP-1; MGC111427; MGC126532; MGC138156
IgM
Caprylic acid/ammonium sulfate precipitation
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.