Learn More
YWHAB Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10936-20UL
 View more versions of this product
Product Code. 18016865
  • 145.00€ / 20µL
Description
The YWHAB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human YWHAB. This antibody recognizes human, mouse, rat antigen. The YWHAB antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
YWHAB
1000 μg/mL
14 3 3 protein beta/alpha, 14 3 3,, GW128, HS1, KCIP 1, Protein 1054, YWHAB
20 μL
7529
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa