missing translation for 'onlineSavingsMsg'
Learn More

YTHDC1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

213.00€ - 507.00€

Specifications

Antigen YTHDC1
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231306
View Documents
Novus Biologicals
NBP3-33486-100ul
100 μL
507.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227741
View Documents
Novus Biologicals
NBP3-33486-20ul
20 μL
213.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

YTHDC1 Monoclonal antibody specifically detects YTHDC1 in Human samples. It is validated for ELISA,Western Blot
Specifications

Specifications

YTHDC1
ELISA, Western Blot
Unconjugated
Rabbit
Human
KIAA1966splicing factor YT521-B, Putative splicing factor YT521, YT521-B, YT521YTH domain-containing protein 1, YTH domain containing 1
Recombinant fusion protein containing a sequence corresponding to amino acids 597-736 of mouse YTHDC1 (NP_808348.2).,, Sequence:, SYNDYVREFHNMGPPPPWQGMPPYPGIEQPPHHPYYQHHAPPPQAHPPYSGHHPVPHEARYRDKRVHDYDMRVDDFLRRTQAVVSGRRSRPRERDRERERDRPRDNRRDRERDRGRDRERERERICDRDRDRGERGRYRR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol, 0.05% BSA
91746
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.