missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.
Specifications
Specifications
| Antigen | YME1L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2B7E4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | YME1L1 |
| Gene Accession No. | Q925S8, Q96TA2 |
| Gene Alias | FTSH, FTSH1, Meg 4, MEG4, PAMP, YME1 like 1 (S. cerevisiae), YME1 like protein 1, YME1L, YME1L1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?