Learn More
YIPF6 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33490-20ul
This item is not returnable.
View return policy
Description
YIPF6 Monoclonal antibody specifically detects YIPF6 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| YIPF6 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Yip1 domain family, member 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-83 of mouse YIPF6 (NP_997516.1).,, Sequence:, MAEAEDSPGEQEAAASKPLFAGLSDVSISQDIPIEGEITIPSRARAQEHDSSTLNESIRRTIMRDLKAVGRKFMHVLYPRKSN | |
| 20 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 286451 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |