missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YIPF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97649-50ul
This item is not returnable.
View return policy
Description
YIPF4 Polyclonal antibody specifically detects YIPF4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| YIPF4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| FinGER4, MGC11061, protein YIPF4, putative protein product of Nbla11189, Yip1 domain family, member 4, YIP1 family member 4 | |
| Produced in rabbits immunized with E. coli-derived Human YIPF4 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84272 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction