missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YIPF4 Antibody, CoraFluor™ 1, Novus Biologicals™
Description
YIPF4 Polyclonal antibody specifically detects YIPF4 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | YIPF4 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | FinGER4, MGC11061, protein YIPF4, putative protein product of Nbla11189, Yip1 domain family, member 4, YIP1 family member 4 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human YIPF4 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?