Learn More
Witeg™ Yellow Safety Washing Bottles
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Ethyl Acetate
Methanol
Methylene chloride
Unit Size:
Each
Description
- Safety washing bottles
- PE
- With safety-delivery jet
- Various capacities
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Yellow |
| Includes | With safety spray |
| Material | LDPE |
| No. per Pack | 1Each |
| Packaging | 1Each |
| Quantity | 1Each |
| Product Type | Wash Bottle |
| Chemical Label | Methylene chloride |