missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YB1 Mouse anti-Human, Clone: YBX1/2430, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | YB1 |
|---|---|
| Clone | YBX1/2430 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Description
YB1 Monoclonal specifically detects YB1 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| YB1 | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control | |
| 10 mM PBS with 0.05% BSA | |
| 4904 | |
| IgG2b κ | |
| Protein A or G purified |
| YBX1/2430 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1 | |
| Recombinant human full-length YB1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title