missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
252.00€ - 307.65€
Specifications
| Antigen | YB1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632396
|
Novus Biologicals
NBP2-98908-100ul |
100 μg |
326.00€ 307.65€ / 100µL Save 18.35€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633866
|
Novus Biologicals
NBP2-98908-50ul |
50 μg |
252.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
YB1 Polyclonal antibody specifically detects YB1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| YB1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control | |
| 0.2 um filtered solution in PBS | |
| 4904 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human YB1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title