missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YB1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98908R
This item is not returnable.
View return policy
Description
YB1 Polyclonal specifically detects YB1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| YB1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| DyLight 550 | |
| BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human YB1. | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control | |
| 4904 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction